Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178803.2 | NP_848918.1 | 4 | 875 | ||
GenBank mRNA | AB221647.1 | - | 4 | 526 | ||
AF090326.1 | - | 610 | ||||
AK029011.1 | - | 4 | 877 | |||
AK045838.1 | - | 4 | 641 | |||
AK045987.1 | - | 4 | 641 | |||
AK155272.1 | - | 4 | 607 | |||
BC025467.1 | - | 4 | 1333 | 5351959 | ||
BC031376.1 | - | 5 | 736 | 5356616 | ||
BC057681.1 | - | 4 | 655 | 3499299 |