Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035145.1 | - | 2, 3 | 331 | ||
AK042326.1 | - | 2, 3 | 335 | |||
AK043506.1 | - | 2, 3 | 325 | |||
AK078626.1 | - | 2, 3 | 331 | |||
AK079602.1 | - | 2, 3 | 306 | |||
AK128913.1 | - | 2, 3 | 268 | |||
AK146286.1 | - | 2, 3 | 324 | |||
AK150233.1 | - | 2, 3 | 323 | |||
AK150250.1 | - | 2, 3 | 336 | |||
AK150920.1 | - | 2, 3 | 323 | |||
AK150986.1 | - | 2, 3 | 305 | |||
AK151763.1 | - | 2, 3 | 323 | |||
AK151826.1 | - | 2, 3 | 305 | |||
AK153128.1 | - | 2, 3 | 305 | |||
AK160060.1 | - | 2, 3 | 325 | |||
AK163452.1 | - | 331 | ||||
AK165032.1 | - | 2, 3 | 569 | |||
BC005476.1 | - | 2, 3 | 313 | 3600875 | ||
S71509.1 | - | 2, 3 | 290 | |||
U07235.1 | - | 2, 3 | 330 |