Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ampd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276301.1 | NP_001263230.1 | 2 | 352 | ||
NM_009667.3 | NP_033797.2 | 2 | 471 | |||
GenBank mRNA | AK133465.1 | - | 2 | 338 | ||
AK171276.1 | - | 2 | 288 | |||
BC040366.1 | - | 2 | 471 | 4503933 | ||
BC056380.1 | - | 2 | 313 | 6851948 | ||
D85596.1 | - | 2 | 350 |