Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009537.3 | NP_033563.2 | 5 | 1479 | ||
GenBank mRNA | AK076149.1 | - | 6 | 1092 | ||
AK166542.1 | - | 6 | 817 | |||
BC055899.1 | - | 5 | 1457 | 3156776 | ||
M73963.1 | - | 5 | 1479 | |||
M74590.1 | - | 5 | 1419 |