Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zap70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289765.1 | NP_001276694.1 | 5 | 931 | ||
NM_001289766.1 | NP_001276695.1 | 5 | 896 | |||
NM_009539.3 | NP_033565.2 | 5 | 888 | |||
GenBank mRNA | AK039883.1 | - | 5 | 825 | ||
AK089056.1 | - | 5 | 861 | |||
BC029727.1 | - | 5 | 846 | 4925739 | ||
U04379.1 | - | 5 | 842 | |||
U77667.1 | - | 4 | 734 |