Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009551.5 | NP_033577.1 | 6 | 1476 | ||
GenBank mRNA | AF062071.1 | - | 5 | 1068 | ||
AK076397.1 | - | 6 | 1477 | |||
AK131911.1 | - | 6 | 1472 | |||
BC061693.1 | - | 2 | 4220 | |||
BC080270.1 | - | 6 | 1457 | |||
BC107566.1 | - | 6 | 1040 | 5389770 |