Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024846.1 | NP_001020017.1 | 5 | 2792 | ||
NM_009562.2 | NP_033588.2 | 4 | 2692 | |||
GenBank mRNA | AK028351.1 | - | 5 | 2795 | ||
AK048619.1 | - | 4 | 2694 | |||
AK154767.1 | - | 4 | 2683 | |||
AK165654.1 | - | 4 | 2673 | |||
AK167091.1 | - | 4 | 2710 | |||
BC022935.1 | - | 4 | 2810 | 3497847 | ||
BC065692.1 | - | 2 | 2896 | 6826605 | ||
L36315.1 | - | 1646 | ||||
Z67747.1 | - | 1 | 1345 |