Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 16 | 4761 | ||
NM_009582.4 | NP_033608.3 | 14 | 4585 | |||
GenBank mRNA | AK029882.1 | - | 14 | 4394 |