Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009602.4 | NP_033732.2 | 6 | 2512 | ||
GenBank mRNA | AK049722.1 | - | 6 | 2485 | ||
AK051742.1 | - | 6 | 2494 | |||
BC065103.1 | - | 6 | 2465 | 6838257 |