Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009605.4 | NP_033735.3 | 2, 3 | 336 | ||
GenBank mRNA | AK003138.1 | - | 2, 3 | 254 | ||
AK041214.1 | - | 2, 3 | 286 | |||
AK134112.1 | - | 2, 3 | 336 | |||
AX767957.1 | - | 2, 3 | 267 | |||
AY749429.1 | - | 1, 2 | 222 | |||
AY754346.1 | - | 1, 2 | 222 | |||
BC028770.1 | - | 2, 3 | 251 | 1329799 | ||
U37222.1 | - | 2, 3 | 267 |