Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301281.1 | NP_001288210.1 | 5 | 1767 | ||
NM_009642.4 | NP_033772.2 | 5 | 1814 | |||
GenBank mRNA | AK036598.1 | - | 5 | 1743 | ||
AK051014.1 | - | 1 | 232 | |||
BC046820.1 | - | 5 | 1800 | 6400135 |