Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301211.1 | NP_001288140.1 | 2 | 457 | ||
NM_009677.6 | NP_033807.2 | 2 | 457 | |||
GenBank mRNA | AK036387.1 | - | 2 | 400 | ||
AK145821.1 | - | 2 | 379 | |||
BC006691.1 | - | 2 | 134 | |||
BC052703.1 | - | 2 | 330 | 5698075 | ||
BC054535.1 | - | 2 | 330 | 5698075 | ||
X54424.1 | - | 2 | 379 |