Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xiap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301639.1 | NP_001288568.1 | 2 | 687 | ||
NM_001301641.1 | NP_001288570.1 | 2 | 474 | |||
NM_009688.3 | NP_033818.2 | 3 | 559 | |||
GenBank mRNA | AK147447.1 | - | 2 | 688 | ||
BC138528.1 | - | 1 | 482 | 8861548 | ||
BC145861.1 | - | 1 | 482 | 40131164 | ||
U36842.1 | - | 2 | 391 | |||
U88990.1 | - | 1 | 851 |