Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277944.1 | NP_001264873.1 | 4 | 373 | ||
GenBank mRNA | AA059961.1 | - | 1 | 19 | ||
AK146468.1 | - | 3 | 174 | |||
BC024697.1 | - | 4 | 373 | 4195301 | ||
BG085960.2 | - | 3 | 216 | |||
CB949564.1 | - | 3 | 279 | |||
Z15090.1 | - | 3 | 174 |