Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010261.1 | - | 4 | 299 | ||
AK131624.1 | - | 4 | 299 | |||
AK134921.1 | - | 4 | 299 | |||
AK148747.1 | - | 4 | 300 | |||
AK149111.1 | - | 4 | 301 | |||
AK149568.1 | - | 4 | 281 | |||
AK150834.1 | - | 4 | 301 | |||
AK159105.1 | - | 4 | 299 | |||
AK159424.1 | - | 4 | 301 | |||
AK159517.1 | - | 4 | 299 | |||
BC028816.1 | - | 4 | 278 | 5136415 | ||
BC044785.1 | - | 3 | 270 | |||
BC083351.1 | - | 3 | 268 | 6818357 | ||
CT010212.1 | - | 3 | 233 | |||
CT010356.1 | - | 3 | 233 | |||
DN173921.1 | - | 3 | 401 | |||
M12414.1 | - | 3 | 233 | |||
M73490.1 | - | 2 | 160 |