Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009739.3 | NP_033869.1 | 4 | 575 | ||
GenBank mRNA | AF043070.1 | - | 3 | 416 | ||
AK075602.1 | - | 4 | 501 | |||
AK145479.1 | - | 4 | 546 | |||
AK150607.1 | - | 4 | 567 | |||
AK150723.1 | - | 4 | 567 | |||
AK167519.1 | - | 4 | 567 | |||
BC046595.1 | - | 4 | 545 | 6313106 |