Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000164.3 | NP_000155.1 | 10 | 986 | ||
NM_001308418.1 | NP_001295347.1 | 9 | 878 | |||
XM_011526709.2 | XP_011525011.1 | 10 | 1043 | |||
XM_011526710.2 | XP_011525012.1 | 11 | 1124 | |||
XM_011526711.2 | XP_011525013.1 | 9 | 935 | |||
XM_011526713.2 | XP_011525015.1 | 8 | 794 | |||
XM_011526714.2 | XP_011525016.1 | 7 | 719 | |||
XM_011526715.2 | XP_011525017.1 | 7 | 729 | |||
XM_011526716.2 | XP_011525018.1 | 10 | 1003 | |||
XM_017026584.1 | XP_016882073.1 | 4 | 506 | |||
XM_017026585.1 | XP_016882074.1 | 3 | 485 | |||
XM_017026586.1 | XP_016882075.1 | 4 | 367 | |||
XM_017026587.1 | XP_016882076.1 | 3 | 485 | |||
GenBank mRNA | BC093723.1 | - | 10 | 973 | 7939568 | |
BC095494.1 | - | 9 | 885 | |||
BC101673.1 | - | 10 | 973 | 8069179 | ||
BC143300.1 | - | 9 | 866 | 9051804 | ||
S79852.1 | - | 10 | 981 | |||
U39231.1 | - | 10 | 986 |
Set Membership: |
G-protein coupled receptor |