Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009787.2 | NP_033917.2 | 3 | 531 | ||
GenBank mRNA | AK028292.1 | - | 3 | 381 | ||
AK146288.1 | - | 3 | 364 | |||
AK148801.1 | - | 3 | 450 | |||
AK150566.1 | - | 3 | 412 | |||
AK161534.1 | - | 3 | 369 | |||
AK168447.1 | - | 3 | 334 | |||
AK169387.1 | - | 3 | 376 | |||
BC066857.1 | - | 2 | 130 | |||
BC141078.1 | - | 3 | 427 | 9055690 | ||
J05186.1 | - | 3 | 414 | |||
Y00884.1 | - | 2 | 237 |