Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009791.4 | NP_033921.3 | 18, 19 | 7931 | ||
GenBank mRNA | AF062378.1 | - | 1, 2 | 191 | ||
AF533752.1 | - | 18, 19 | 7909 | |||
AK048277.1 | - | 1, 2 | 743 | |||
AY971958.1 | - | 17, 18 | 3977 |