Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110794.1 | NP_001104264.1 | 5 | 624 | ||
NM_009674.3 | NP_033804.2 | 5 | 739 | |||
GenBank mRNA | AK032013.1 | - | 5 | 515 | ||
AK077579.1 | - | 5 | 432 | |||
AK134589.1 | - | 5 | 443 | |||
AK149654.1 | - | 5 | 431 | |||
AK151438.1 | - | 5 | 433 | |||
AK161613.1 | - | 5 | 433 | |||
AK167588.1 | - | 5 | 407 | |||
AK169199.1 | - | 5 | 406 | |||
BC008997.1 | - | 5 | 423 | 3157926 | ||
L13129.1 | - | 5 | 426 |