Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037737.2 | NP_001032826.1 | 2, 3 | 291 | ||
NM_009709.4 | NP_033839.2 | 2, 3 | 291 | |||
GenBank mRNA | AK028546.1 | - | 2, 3 | 278 | ||
AK040475.1 | - | 2, 3 | 293 | |||
AK141362.1 | - | 2, 3 | 162 | |||
AK145415.1 | - | 2, 3 | 246 | |||
AK152473.1 | - | 2, 3 | 273 | |||
AK153355.1 | - | 2, 3 | 273 | |||
BC012870.1 | - | 2, 3 | 253 | 3964932 | ||
CJ183661.1 | - | 2, 3 | 291 | |||
U10325.1 | - | 2, 3 | 155 | |||
U14333.1 | - | 2, 3 | 144 |