Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Artn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284191.1 | NP_001271120.1 | 6 | 1645 | ||
NM_001284192.1 | NP_001271121.1 | 5 | 2135 | |||
NM_001284193.1 | NP_001271122.1 | 5 | 2131 | |||
NM_009711.4 | NP_033841.1 | 5 | 1987 | |||
GenBank mRNA | AK015393.1 | - | 6 | 1353 | ||
AK053914.1 | - | 5 | 1988 | |||
AK145190.1 | - | 5 | 1856 | |||
AK156507.1 | - | 5 | 1889 | |||
AK166013.1 | - | 4 | 1732 | |||
BC104328.1 | - | 2 | 865 | |||
BC104329.1 | - | 3 | 545 |