Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038999.2 | NP_001034088.1 | 3 | 429 | ||
NM_001284345.1 | NP_001271274.1 | 3 | 429 | |||
NM_009727.3 | NP_033857.1 | 3 | 429 | |||
GenBank mRNA | AK045367.1 | - | 3 | 410 | ||
AK076388.1 | - | 3 | 368 | |||
AK141559.1 | - | 3 | 364 | |||
AK151709.1 | - | 3 | 430 | |||
AK220560.1 | - | 3 | 339 | |||
BC129872.1 | - | 3 | 204 | 40110214 | ||
U75321.1 | - | 3 | 208 |