Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009748.2 | NP_033878.1 | 4 | 493 | ||
GenBank mRNA | AK078796.1 | - | 4 | 489 | ||
BC005572.1 | - | 4 | 493 | 3967968 |