Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243211.1 | NP_001230140.1 | 5 | 603 | ||
NM_001562.3 | NP_001553.1 | 6 | 615 | |||
XM_011542805.1 | XP_011541107.1 | 5 | 611 | |||
XM_011542806.2 | XP_011541108.1 | 6 | 619 | |||
GenBank mRNA | AF077611.1 | - | 5 | 395 | ||
AF380360.1 | - | 6 | 447 | |||
AY044641.1 | - | 6 | 492 | |||
AY266351.1 | - | 5 | 560 | |||
BC007007.1 | - | 6 | 590 | 3686124 | ||
BC007461.1 | - | 6 | 615 | 3681009 | ||
CR541973.1 | - | 5 | 395 | |||
CR542001.1 | - | 5 | 395 | |||
D49950.1 | - | 6 | 572 | |||
U90434.1 | - | 5 | 392 |