Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009780.2 | NP_033910.2 | 14 | 1810 | ||
GenBank mRNA | AK128974.1 | - | 2 | 236 | ||
AK157954.1 | - | 14 | 1796 | |||
AK158256.1 | - | 14 | 1796 | |||
AK158475.1 | - | 14 | 1796 | |||
AK171644.1 | - | 14 | 1796 | |||
AK172637.1 | - | 14 | 1798 | |||
BC039141.1 | - | 14 | 1773 | |||
BC055297.1 | - | 14 | 1780 | |||
BC067394.1 | - | 14 | 1780 | 30107263 | ||
BC067409.1 | - | 14 | 1780 | 30098613 | ||
BC150743.1 | - | 14 | 1760 | 9087654 | ||
BC158005.1 | - | 14 | 1759 | 9007313 | ||
D10084.1 | - | 340 | ||||
M11729.1 | - | 14 | 1795 |