Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009787.2 | NP_033917.2 | 3 | 639 | ||
GenBank mRNA | AK028292.1 | - | 3 | 489 | ||
AK146288.1 | - | 3 | 472 | |||
AK148801.1 | - | 3 | 558 | |||
AK150566.1 | - | 3 | 520 | |||
AK161534.1 | - | 3 | 477 | |||
AK168447.1 | - | 3 | 442 | |||
AK169387.1 | - | 3 | 484 | |||
BC066857.1 | - | 2 | 238 | |||
BC141078.1 | - | 3 | 535 | 9055690 | ||
J05186.1 | - | 3 | 522 | |||
Y00884.1 | - | 2 | 345 |