Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LALBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002289.2 | NP_002280.1 | 3, 4 | 389 | ||
XM_006719395.3 | XP_006719458.1 | 4, 5 | 467 | |||
GenBank mRNA | BC069103.1 | - | 3, 4 | 389 | 7216918 | |
BC112316.1 | - | 3, 4 | 385 | 8327784 | ||
BC112318.1 | - | 3, 4 | 385 | 8327786 | ||
BX281629.1 | - | 1, 2 | 65 | |||
CR541987.1 | - | 3, 4 | 363 | |||
CR542017.1 | - | 3, 4 | 363 | |||
J00270.1 | - | 3, 4 | 363 |