Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009672.3 | NP_033802.2 | 4, 5 | 688 | ||
GenBank mRNA | BC062899.1 | - | 4, 5 | 603 | 6415168 | |
U73478.1 | - | 4, 5 | 555 |