Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009673.2 | NP_033803.1 | 4 | 510 | ||
GenBank mRNA | AK147740.1 | - | 4 | 289 | ||
AK152185.1 | - | 4 | 345 | |||
AK153388.1 | - | 4 | 291 | |||
AK153476.1 | - | 4 | 274 | |||
BC003716.1 | - | 4 | 301 | 3488901 | ||
CX218962.1 | - | 4 | 466 | |||
D63423.1 | - | 4 | 248 | |||
U29396.1 | - | 4 | 267 |