Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042558.1 | NP_001036023.1 | 14, 15 | 2716 | ||
NM_001282947.1 | NP_001269876.1 | 14, 15 | 2640 | |||
NM_009684.2 | NP_033814.2 | 14, 15 | 2673 | |||
GenBank mRNA | AF064071.1 | - | 14, 15 | 2600 | ||
AK220340.1 | - | 14, 15 | 2712 | |||
BC083191.1 | - | 14, 15 | 2590 | |||
BC131683.1 | - | 14, 15 | 2232 | 40111198 | ||
BC131684.1 | - | 14, 15 | 2232 | 40111199 |