Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159645.1 | NP_001153117.1 | 3, 4 | 344 | ||
NM_009703.2 | NP_033833.1 | 3, 4 | 344 | |||
GenBank mRNA | AB158252.1 | - | 2, 3 | 269 | ||
AK004741.1 | - | 2, 3 | 1623 | |||
AK010060.1 | - | 1, 2 | 95 | |||
AK020547.1 | - | 3, 4 | 310 | |||
AK034861.1 | - | 3, 4 | 339 | |||
AK039044.1 | - | 3, 4 | 310 | |||
AK085823.1 | - | 3, 4 | 310 | |||
AK133797.1 | - | 2, 3 | 1630 | |||
AK135358.1 | - | 3, 4 | 310 | |||
AK163567.1 | - | 3, 4 | 313 | |||
AK169757.1 | - | 3, 4 | 325 | |||
BB857915.1 | - | 3, 4 | 344 | |||
BC004757.1 | - | 3, 4 | 300 | 3154293 |