Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287530.1 | NP_001274459.1 | 13 | 3921 | ||
NM_009707.4 | NP_033837.2 | 13 | 5289 | |||
NM_178754.3 | NP_848869.1 | 13 | 3832 | |||
GenBank mRNA | AF012273.2 | - | 4211 | |||
AF177664.1 | - | 4350 | ||||
AK077996.1 | - | 13 | 3794 | |||
AK079651.1 | - | 15 | 3884 | |||
AK168658.1 | - | 1 | 1822 |