Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009739.3 | NP_033869.1 | 12 | 1494 | ||
GenBank mRNA | AF043070.1 | - | 11 | 1335 | ||
AK075602.1 | - | 12 | 1420 | |||
AK145479.1 | - | 12 | 1465 | |||
AK150607.1 | - | 12 | 1486 | |||
AK150723.1 | - | 12 | 1486 | |||
AK167519.1 | - | 12 | 1486 | |||
AK203967.1 | - | 1 | 80 | |||
BC004077.1 | - | 9 | 922 | |||
BC046595.1 | - | 12 | 1464 | 6313106 |