Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009745.2 | NP_033875.2 | 6 | 1003 | ||
GenBank mRNA | AJ011145.1 | - | 6 | 883 | ||
AK131643.1 | - | 6 | 899 | |||
AK155076.1 | - | 6 | 885 | |||
AK219291.1 | - | 1 | 180 | |||
BC011471.1 | - | 6 | 980 | 3660754 |