Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnip3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009761.3 | NP_033891.1 | 6 | 2224 | ||
GenBank mRNA | AK004667.1 | - | 6 | 2170 | ||
AK018668.1 | - | 6 | 2176 | |||
AK139950.1 | - | 6 | 2169 | |||
AK155040.1 | - | 6 | 2194 | |||
AK168482.1 | - | 6 | 2227 | |||
AK169391.1 | - | 6 | 2227 | |||
AY057069.1 | - | 7 | 2243 |