Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011710.1 | - | 3 | 150 | ||
AK146363.1 | - | 3 | 160 | |||
AK164321.1 | - | 3 | 126 | |||
AK165998.1 | - | 3 | 137 | |||
AK168842.1 | - | 3 | 158 | |||
BC002022.1 | - | 3 | 146 | 3491126 | ||
BY070840.1 | - | 3 | 186 | |||
BY265671.1 | - | 3 | 196 | |||
Y13087.1 | - | 2 | 114 |