Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190333.1 | NP_001177262.1 | 3 | 494 | ||
NM_001190334.1 | NP_001177263.1 | 4 | 366 | |||
NM_001190335.1 | NP_001177264.1 | 3 | 275 | |||
NM_001190336.1 | NP_001177265.1 | 3 | 251 | |||
NM_001190337.1 | NP_001177266.1 | 2 | 160 | |||
NM_001190338.1 | NP_001177267.1 | 2 | 247 | |||
NM_009835.4 | NP_033965.1 | 6 | 696 | |||
GenBank mRNA | AB009369.1 | - | 2 | 250 | ||
AB016031.1 | - | 3 | 325 | |||
AK089712.1 | - | 6 | 697 | |||
AK154413.1 | - | 5 | 1069 | |||
BC100756.1 | - | 4 | 362 | 40046797 | ||
BC100757.1 | - | 3 | 271 | 40046796 | ||
BC100758.1 | - | 3 | 247 | 40046794 | ||
BC105669.1 | - | 2 | 153 | 40046795 |