Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043317.2 | NP_001036782.1 | 5 | 533 | ||
NM_009845.3 | NP_033975.3 | 3 | 408 | |||
GenBank mRNA | AF102134.1 | - | 2 | 177 | ||
AF115400.1 | - | 2 | 177 | |||
AF115401.1 | - | 2 | 177 | |||
AJ250676.1 | - | 2 | 338 | |||
AJ250677.1 | - | 2 | 325 | |||
AJ250678.1 | - | 2 | 307 | |||
AJ250679.1 | - | 2 | 270 | |||
AJ250680.1 | - | 3 | 239 | |||
AJ250682.1 | - | 2 | 270 | |||
AK041230.1 | - | 5 | 542 | |||
AK143835.1 | - | 5 | 471 | |||
AK156735.1 | - | 3 | 407 | |||
AK171589.1 | - | 2 | 214 | |||
AK171990.1 | - | 2 | 528 | |||
AK172301.1 | - | 3 | 364 | |||
BC046957.1 | - | 5 | 459 | |||
BC150807.1 | - | 4 | 492 | 9087718 | ||
L02844.1 | - | 4 | 679 | |||
L16928.1 | - | 4 | 674 |