Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009849.2 | NP_033979.2 | 2 | 293 | ||
GenBank mRNA | AF042811.1 | - | 2 | 217 | ||
AK002553.1 | - | 2 | 265 | |||
AK172281.1 | - | 2 | 241 | |||
AY376711.1 | - | 2 | 226 | |||
BC011241.1 | - | 2 | 225 | 3582550 | ||
BY237099.1 | - | 2 | 294 | |||
U91511.1 | - | 2 | 260 |