Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037801.2 | NP_001032890.1 | 3 | 387 | ||
NM_009852.3 | NP_033982.3 | 3 | 387 | |||
GenBank mRNA | AK030822.1 | - | 3 | 357 | ||
AK030823.1 | - | 3 | 388 | |||
AK088050.1 | - | 3 | 365 | |||
BC014294.1 | - | 3 | 393 | 4187689 | ||
U12434.1 | - | 3 | 185 | |||
U35370.1 | - | 3 | 303 | |||
U37543.1 | - | 3 | 344 | |||
U37544.1 | - | 3 | 344 |