Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009872.3 | NP_034002.2 | 1 | 312 | ||
GenBank mRNA | BB622496.1 | - | 1 | 314 | ||
BC100712.1 | - | 1 | 241 | |||
BC100714.1 | - | 1 | 237 | |||
BC100715.1 | - | 1 | 237 | |||
BC103550.1 | - | 1 | 240 |