Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009873.2 | NP_034003.1 | 1, 2 | 296 | ||
GenBank mRNA | AF132482.1 | - | 1, 2 | 127 | ||
AF132483.1 | - | 1, 2 | 296 | |||
AK078973.1 | - | 1, 2 | 407 | |||
AK154197.1 | - | 2, 3 | 569 | |||
BC119360.1 | - | 1, 2 | 285 | 8734109 | ||
BC120595.1 | - | 1, 2 | 285 | 40129518 |