Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009888.3 | NP_034018.2 | 2 | 264 | ||
GenBank mRNA | AK052688.1 | - | 2 | 264 | ||
AK087707.1 | - | 2 | 173 | |||
BB661130.1 | - | 2 | 264 | |||
BC066092.1 | - | 2 | 133 | 6842070 | ||
M12660.1 | - | 2 | 175 |