Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF047838.1 | - | 582 | |||
AF052746.1 | - | 4, 5 | 611 | |||
AK032511.1 | - | 4, 5 | 581 | |||
AK087213.1 | - | 4, 5 | 583 | |||
AK142211.1 | - | 4, 5 | 583 | |||
AK161805.1 | - | 4, 5 | 584 | |||
BC010260.1 | - | 4, 5 | 820 | 3710379 | ||
BC132342.1 | - | 4, 5 | 585 | 40130619 | ||
BC145057.1 | - | 585 | 9053605 |