Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009794.3 | NP_033924.2 | 4 | 654 | ||
GenBank mRNA | AF015038.1 | - | 4 | 600 | ||
AK052141.1 | - | 4 | 629 | |||
AK053832.1 | - | 5 | 914 | |||
AK078787.1 | - | 1 | 465 | |||
AK150695.1 | - | 4 | 628 | |||
AK152094.1 | - | 4 | 628 | |||
AK152790.1 | - | 4 | 628 | |||
AK152994.1 | - | 4 | 628 | |||
AK154855.1 | - | 4 | 668 | |||
AK184788.1 | - | 51 | ||||
BC054726.1 | - | 4 | 654 | 5703563 | ||
D38117.1 | - | 4 | 695 | |||
Y10139.1 | - | 4 | 536 |