Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009814.3 | NP_033944.2 | 1, 2 | 411 | ||
GenBank mRNA | AF068244.1 | - | 1, 2 | 356 | ||
AI114962.1 | - | 1, 2 | 411 | |||
AW106087.1 | - | 1, 2 | 356 | |||
BC092229.1 | - | 1, 2 | 324 | 30603069 | ||
BC137668.1 | - | 1, 2 | 320 | 8860687 | ||
BC140959.1 | - | 1, 2 | 320 | 9055571 | ||
U91483.1 | - | 1, 2 | 230 |