Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009831.2 | NP_033961.1 | 2, 3 | 555 | ||
GenBank mRNA | AK128957.1 | - | 2, 3 | 543 | ||
AK143114.1 | - | 2, 3 | 546 | |||
AK147026.1 | - | 2, 3 | 543 | |||
AK153392.1 | - | 2, 3 | 487 | |||
AK159429.1 | - | 2, 3 | 485 | |||
AK159831.1 | - | 2, 3 | 490 | |||
AK160411.1 | - | 2, 3 | 530 | |||
AK167052.1 | - | 2, 3 | 487 | |||
AK167486.1 | - | 2, 3 | 482 | |||
AK170397.1 | - | 2, 3 | 543 | |||
AK178990.1 | - | 1, 2 | 68 | |||
AK186078.1 | - | 1 | 18 | |||
AK219604.1 | - | 1 | 20 | |||
BC005534.1 | - | 1, 2 | 205 | |||
BC006609.1 | - | 2, 3 | 439 | |||
L49507.1 | - | 2, 3 | 498 | |||
Z37110.1 | - | 2, 3 | 498 |