Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ250676.1 | - | 2 | 386 | ||
AJ250677.1 | - | 2 | 373 | |||
AJ250678.1 | - | 2 | 355 | |||
AJ250679.1 | - | 2 | 318 | |||
AJ250680.1 | - | 3 | 287 | |||
AJ250681.1 | - | 218 | ||||
AJ250682.1 | - | 2 | 318 | |||
AJ250684.1 | - | 3 | 132 | |||
L16928.1 | - | 4 | 722 |