Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009858.2 | NP_033988.1 | 2 | 239 | ||
GenBank mRNA | AK087944.1 | - | 2 | 216 | ||
AK088372.1 | - | 2 | 264 | |||
AK088649.1 | - | 2 | 263 | |||
AK154055.1 | - | 2 | 219 | |||
AK162993.1 | - | 2 | 228 | |||
AK169581.1 | - | 2 | 240 | |||
M16799.1 | - | 2 | 232 | |||
M17534.1 | - | 2 | 239 | |||
M19504.1 | - | 2 | 227 | |||
U34882.1 | - | 2 | 359 | |||
X07698.1 | - | 2 | 243 |